siRNA Details
| Gene Symbol: Capn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109761.2 | NP_001103231.1 | 17 | 1877 | ||
| NM_001177799.1 | NP_001171270.1 | 17 | 2144 | |||
| NM_007601.3 | NP_031627.2 | 20 | 2420 | |||
| GenBank mRNA | AF091998.1 | - | 17 | 1841 | ||
| AF127766.2 | - | 17 | 1864 | |||
| AK049994.1 | - | 7 | 306 | |||
| AK079673.1 | - | 2 | 96 | |||
| AK088613.1 | - | 5 | 631 | |||
| BC090661.1 | - | 17 | 1877 | 30681055 | ||
| BC139790.1 | - | 20 | 2140 | 40047690 | ||
| X92523.1 | - | 20 | 2140 | |||

Capn3
Genomic Map