siRNA Details
| Gene Symbol: Cav3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007617.3 | NP_031643.1 | 1 | 361 | ||
| GenBank mRNA | BC024383.1 | - | 1 | 371 | 4988598 | |
| CJ057698.1 | - | 1 | 138 | |||
| U36579.1 | - | 1 | 175 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cav3| Gene Symbol: Cav3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007617.3 | NP_031643.1 | 1 | 361 | ||
| GenBank mRNA | BC024383.1 | - | 1 | 371 | 4988598 | |
| CJ057698.1 | - | 1 | 138 | |||
| U36579.1 | - | 1 | 175 | |||