siRNA Details
| Gene Symbol: Cdk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007668.3 | NP_031694.1 | 6 | 412 | ||
| GenBank mRNA | AK051790.1 | - | 6 | 412 | ||
| BC052007.1 | - | 6 | 386 | 5706932 | ||
| D29678.1 | - | 6 | 421 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk5| Gene Symbol: Cdk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007668.3 | NP_031694.1 | 6 | 412 | ||
| GenBank mRNA | AK051790.1 | - | 6 | 412 | ||
| BC052007.1 | - | 6 | 386 | 5706932 | ||
| D29678.1 | - | 6 | 421 | |||