siRNA Details
| Gene Symbol: Cdk5r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009872.3 | NP_034002.2 | 1 | 316 | ||
| GenBank mRNA | BB622496.1 | - | 1 | 318 | ||
| BC100712.1 | - | 1 | 245 | |||
| BC100714.1 | - | 1 | 241 | |||
| BC100715.1 | - | 1 | 241 | |||
| BC103550.1 | - | 1 | 244 | |||

Cdk5r2
Genomic Map