siRNA Details
| Gene Symbol: Cdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007673.3 | NP_031699.2 | 3 | 1962 | ||
| GenBank mRNA | AK018629.1 | - | 1 | 986 | ||
| AK049566.1 | - | 3 | 1963 | |||
| AK049574.1 | - | 3 | 1835 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdx2| Gene Symbol: Cdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007673.3 | NP_031699.2 | 3 | 1962 | ||
| GenBank mRNA | AK018629.1 | - | 1 | 986 | ||
| AK049566.1 | - | 3 | 1963 | |||
| AK049574.1 | - | 3 | 1835 | |||