siRNA Details
| Gene Symbol: Crygf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027010.2 | NP_081286.1 | 3 | 548 | ||
| GenBank mRNA | AK013953.1 | - | 3 | 549 | ||
| BC056453.1 | - | 3 | 539 | 6824171 | ||
| Gene Symbol: Cryge | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007777.3 | NP_031803.3 | 3 | 562 | ||
| GenBank mRNA | AK014301.1 | - | 3 | 549 | ||
| BC078417.1 | - | 537 | 6824205 | |||
| BC132506.1 | - | 3 | 562 | 40130783 | ||
| BC132508.1 | - | 3 | 562 | 40130785 | ||
| K02584.1 | - | 516 | ||||

Crygf
Genomic Map