siRNA Details
| Gene Symbol: Csn1s2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142688.1 | - | 5, 6 | 198 | ||
| AK164738.1 | - | 5, 6 | 202 | |||
| BC139326.1 | - | 5, 6 | 178 | 8862348 | ||
| D10215.1 | - | 5, 6 | 203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csn1s2a| Gene Symbol: Csn1s2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142688.1 | - | 5, 6 | 198 | ||
| AK164738.1 | - | 5, 6 | 202 | |||
| BC139326.1 | - | 5, 6 | 178 | 8862348 | ||
| D10215.1 | - | 5, 6 | 203 | |||