siRNA Details
| Gene Symbol: Cyp17a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007809.3 | NP_031835.3 | 4 | 867 | ||
| GenBank mRNA | AK132830.1 | - | 4 | 761 | ||
| AK134376.1 | - | 4 | 761 | |||
| BC064793.1 | - | 4 | 841 | 30436278 | ||
| M64863.1 | - | 4 | 740 | |||

Cyp17a1
Genomic Map