siRNA Details
| Gene Symbol: Daf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007827.2 | NP_031853.2 | 3 | 375 | ||
| GenBank mRNA | BC115954.1 | - | 3 | 375 | ||
| L41365.1 | - | 3 | 384 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Daf2| Gene Symbol: Daf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007827.2 | NP_031853.2 | 3 | 375 | ||
| GenBank mRNA | BC115954.1 | - | 3 | 375 | ||
| L41365.1 | - | 3 | 384 | |||