siRNA Details
| Gene Symbol: Dapk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010019.3 | NP_034149.2 | 8 | 842 | ||
| GenBank mRNA | AB018002.1 | - | 8 | 832 | ||
| AF052942.1 | - | 6 | 522 | |||
| BC022165.1 | - | 8 | 811 | 4016911 | ||
| BX527735.1 | - | 4 | 226 | |||
| GU109477.1 | - | 8 | 842 | |||

Dapk2
Genomic Map