siRNA Details
| Gene Symbol: Dbil5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021294.2 | NP_067269.1 | 2 | 442 | ||
| GenBank mRNA | AK006528.1 | - | 2 | 403 | ||
| BC048474.1 | - | 2 | 421 | 6773240 | ||
| CN833562.1 | - | 445 | ||||
| S83465.1 | - | 2 | 429 | |||

Dbil5
Genomic Map