siRNA Details
| Gene Symbol: Ei24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199494.1 | NP_001186423.1 | 11 | 1159 | ||
| NM_007915.5 | NP_031941.1 | 11 | 1043 | |||
| GenBank mRNA | AK078386.1 | - | 1 | 33 | ||
| AK145018.1 | - | 11 | 1121 | |||
| AK147776.1 | - | 11 | 1072 | |||
| AK159927.1 | - | 11 | 1101 | |||
| AK172233.1 | - | 11 | 1123 | |||
| U41751.2 | - | 11 | 1047 | |||

Ei24
Genomic Map