siRNA Details
| Gene Symbol: Elk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007923.2 | NP_031949.2 | 3 | 857 | ||
| GenBank mRNA | AK054075.1 | - | 3 | 1149 | ||
| AK156537.1 | - | 3 | 855 | |||
| BC004798.1 | - | 1 | 102 | |||
| BC041679.1 | - | 1 | 90 | |||
| BC144844.1 | - | 3 | 607 | 9053392 | ||
| BC145795.1 | - | 3 | 607 | 40131098 | ||
| Z36885.1 | - | 4 | 802 | |||

Elk4
Genomic Map