siRNA Details
| Gene Symbol: Emp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146346.1 | NP_001139818.1 | 4 | 514 | ||
| NM_010129.2 | NP_034259.2 | 4 | 485 | |||
| GenBank mRNA | AF011750.1 | - | 4 | 346 | ||
| AK146939.1 | - | 4 | 517 | |||
| BC001999.1 | - | 4 | 330 | 3485948 | ||
| CK022835.1 | - | 4 | 484 | |||
| U87948.1 | - | 4 | 463 | |||

Emp3
Genomic Map