siRNA Details
| Gene Symbol: Epb4.1l4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013512.2 | NP_038540.2 | 10, 11 | 1572 | ||
| GenBank mRNA | AK140485.1 | - | 9, 10 | 1256 | ||
| BC007166.1 | - | 10, 11 | 1296 | 3490061 | ||
| BC011099.1 | - | 9, 10 | 872 | |||
| BC013557.1 | - | 10, 11 | 1296 | 3490326 | ||
| BC089158.1 | - | 9, 10 | 1144 | |||
| D28818.1 | - | 9, 10 | 846 | |||

Epb4.1l4a
Genomic Map