siRNA Details
| Gene Symbol: Eya4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010167.4 | NP_034297.2 | 15 | 1590 | ||
| GenBank mRNA | AJ007997.1 | - | 1 | 17 | ||
| AK041625.1 | - | 15 | 1591 | |||
| BC120898.1 | - | 15 | 1574 | 40061597 | ||
| Y17115.1 | - | 16 | 1701 | |||

Eya4
Genomic Map