siRNA Details
| Gene Symbol: Fadd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010175.5 | NP_034305.1 | 2 | 1538 | ||
| GenBank mRNA | AK031396.1 | - | 2 | 1539 | ||
| AK084808.1 | - | 2 | 1539 | |||
| AK169798.1 | - | 2 | 1566 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fadd| Gene Symbol: Fadd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010175.5 | NP_034305.1 | 2 | 1538 | ||
| GenBank mRNA | AK031396.1 | - | 2 | 1539 | ||
| AK084808.1 | - | 2 | 1539 | |||
| AK169798.1 | - | 2 | 1566 | |||