siRNA Details
Gene Symbol: Gabra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008066.3 | NP_032092.1 | 9 | 1278 | ||
GenBank mRNA | AK039055.1 | - | 9 | 1279 | ||
AK046953.1 | - | 4 | 1412 | |||
AK048165.1 | - | 1176 | ||||
BC115727.1 | - | 9 | 1200 | 40039253 | ||
M86567.1 | - | 9 | 1029 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
