siRNA Details
| Gene Symbol: Gdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010267.3 | NP_034397.1 | 5 | 761 | ||
| GenBank mRNA | AK039016.1 | - | 1 | 1640 | ||
| AK045460.1 | - | 5 | 761 | |||
| AK083814.1 | - | 5 | 762 | |||
| AK139057.1 | - | 5 | 604 | |||
| AK149281.1 | - | 5 | 772 | |||
| BC048177.1 | - | 5 | 737 | 6825756 | ||
| BC051135.1 | - | 5 | 726 | 6532839 | ||
| BC060212.1 | - | 5 | 734 | |||
| Y17850.1 | - | 5 | 749 | |||

Gdap1
Genomic Map