siRNA Details
| Gene Symbol: Gip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008119.2 | NP_032145.2 | 5 | 498 | ||
| GenBank mRNA | AK008308.1 | - | 5 | 498 | ||
| AK008525.1 | - | 4 | 354 | |||
| BC104314.1 | - | 5 | 488 | 40044479 | ||
| BC104315.1 | - | 5 | 488 | 40044480 | ||

Gip
Genomic Map