siRNA Details
| Gene Symbol: Gnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010312.4 | NP_034442.1 | 8 | 815 | ||
| GenBank mRNA | AK146941.1 | - | 8 | 839 | ||
| AK151628.1 | - | 8 | 805 | |||
| BC029077.1 | - | 7 | 655 | 4019292 | ||
| BC059942.1 | - | 8 | 841 | 6529685 | ||
| BC062178.1 | - | 8 | 798 | 6330944 | ||
| U34960.1 | - | 7 | 635 | |||
| U38505.1 | - | 1 | 92 | |||

Gnb2
Genomic Map