siRNA Details
| Gene Symbol: Gngt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038664.2 | NP_001033753.2 | 4 | 541 | ||
| NM_001284393.1 | NP_001271322.1 | 4 | 545 | |||
| NM_001284397.1 | NP_001271326.1 | 4 | 542 | |||
| NM_023121.2 | NP_075610.1 | 4 | 526 | |||
| GenBank mRNA | AA028742.1 | - | 4 | 496 | ||
| AK010554.1 | - | 3 | 1555 | |||
| BC061005.1 | - | 4 | 491 | 30295552 | ||
| BF470980.1 | - | 3 | 355 | |||

Gngt2
Genomic Map