siRNA Details
| Gene Symbol: H2-M9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008205.1 | NP_032231.1 | 2 | 301 | ||
| GenBank mRNA | AF469488.1 | - | 2 | 301 | ||
| AF539595.1 | - | 2 | 301 | |||
| AK080256.1 | - | 2 | 325 | |||
| BC106996.2 | - | 2 | 300 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2-M9| Gene Symbol: H2-M9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008205.1 | NP_032231.1 | 2 | 301 | ||
| GenBank mRNA | AF469488.1 | - | 2 | 301 | ||
| AF539595.1 | - | 2 | 301 | |||
| AK080256.1 | - | 2 | 325 | |||
| BC106996.2 | - | 2 | 300 | |||