siRNA Details
Gene Symbol: H2-DMb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010387.3 | NP_034517.2 | 5, 6 | 990 | ||
GenBank mRNA | AK032745.1 | - | 5, 6 | 991 | ||
BC003718.1 | - | 5, 6 | 930 | 3486844 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: H2-DMb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010387.3 | NP_034517.2 | 5, 6 | 990 | ||
GenBank mRNA | AK032745.1 | - | 5, 6 | 991 | ||
BC003718.1 | - | 5, 6 | 930 | 3486844 |