siRNA Details
| Gene Symbol: Hba-x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010405.4 | NP_034535.1 | 3 | 483 | ||
| GenBank mRNA | AK077590.1 | - | 3 | 482 | ||
| BC051988.1 | - | 3 | 453 | 5711898 | ||
| M13125.1 | - | 2 | 291 | |||
| M26898.1 | - | 3 | 441 | |||

Hba-x
Genomic Map