siRNA Details
| Gene Symbol: Hbb-y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008221.4 | NP_032247.1 | 3 | 513 | ||
| GenBank mRNA | AK011619.1 | - | 3 | 514 | ||
| AK012501.1 | - | 3 | 515 | |||
| AK012897.1 | - | 3 | 514 | |||
| AK077938.1 | - | 1349 | ||||
| BC057014.1 | - | 3 | 490 | 6817810 | ||
| M26897.1 | - | 3 | 475 | |||
| V00857.1 | - | 3 | 393 | |||

Hbb-y
Genomic Map