siRNA Details
| Gene Symbol: Hoxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010450.3 | NP_034580.1 | 1 | 466 | ||
| GenBank mRNA | AK133481.1 | - | 1 | 466 | ||
| BC052232.1 | - | 1 | 444 | |||
| BC119302.1 | - | 1 | 401 | 8734051 | ||
| U20370.1 | - | 1 | 422 | |||

Hoxa11
Genomic Map