siRNA Details
| Gene Symbol: Ifnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010510.1 | NP_034640.1 | 1 | 453 | ||
| GenBank mRNA | BC119395.1 | - | 1 | 453 | 8734144 | |
| BC119397.1 | - | 1 | 453 | 8734146 | ||
| K00020.1 | - | 1 | 453 | |||
| M27321.1 | - | 42 | ||||

Ifnb1
Genomic Map