siRNA Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010560.3 | NP_034690.3 | 16, 17 | 2303 | ||
| GenBank mRNA | AK142637.1 | - | 6, 7 | 764 | ||
| BC058679.1 | - | 16, 17 | 2264 | 6834623 | ||
| BU709006.1 | - | 3, 4 | 219 | |||
| M83336.1 | - | 15, 16 | 2108 | |||
| X62646.1 | - | 15, 16 | 2108 | |||

Il6st
Genomic Map