siRNA Details
| Gene Symbol: Krtap9-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015741.2 | NP_056556.1 | 1 | 360 | ||
| GenBank mRNA | BC116210.2 | - | 1 | 309 | 40091112 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap9-1| Gene Symbol: Krtap9-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015741.2 | NP_056556.1 | 1 | 360 | ||
| GenBank mRNA | BC116210.2 | - | 1 | 309 | 40091112 | |