siRNA Details
| Gene Symbol: Lgals3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145953.1 | NP_001139425.1 | 5, 6 | 764 | ||
| NM_010705.3 | NP_034835.1 | 5, 6 | 737 | |||
| GenBank mRNA | AK008593.1 | - | 4, 5 | 406 | ||
| AK089249.1 | - | 5, 6 | 681 | |||
| AK149783.1 | - | 5, 6 | 711 | |||
| AK155244.1 | - | 6, 7 | 583 | |||
| AK155484.1 | - | 5, 6 | 679 | |||
| AK159536.1 | - | 5, 6 | 705 | |||
| AK170803.1 | - | 5, 6 | 794 | |||
| BC132328.1 | - | 5, 6 | 690 | 40130605 | ||
| BC138790.1 | - | 5, 6 | 690 | 8861812 | ||
| BC145419.1 | - | 5, 6 | 690 | 9053977 | ||
| J03723.1 | - | 3, 4 | 637 | |||
| M33215.1 | - | 3, 4 | 346 | |||
| X16074.1 | - | 5, 6 | 784 | |||
| X16834.1 | - | 5, 6 | 683 | |||

Lgals3
Genomic Map