siRNA Details
| Gene Symbol: Lifr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013584.2 | NP_038612.1 | 16 | 2266 | ||
| GenBank mRNA | AK155524.1 | - | 16 | 2493 | ||
| D26177.1 | - | 16 | 2273 | |||
| S73495.1 | - | 16 | 2273 | |||
| S73496.1 | - | 16 | 2774 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lifr| Gene Symbol: Lifr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013584.2 | NP_038612.1 | 16 | 2266 | ||
| GenBank mRNA | AK155524.1 | - | 16 | 2493 | ||
| D26177.1 | - | 16 | 2273 | |||
| S73495.1 | - | 16 | 2273 | |||
| S73496.1 | - | 16 | 2774 | |||