siRNA Details
| Gene Symbol: Tram2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177409.3 | NP_803128.1 | 7, 8 | 733 | ||
| GenBank mRNA | AK082751.1 | - | 7, 8 | 734 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Tram2
Tram2| Gene Symbol: Tram2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177409.3 | NP_803128.1 | 7, 8 | 733 | ||
| GenBank mRNA | AK082751.1 | - | 7, 8 | 734 | ||
 Back To Top
 Back To Top