siRNA Details
| Gene Symbol: Galnt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134189.2 | NP_598950.2 | 3 | 443 | ||
| GenBank mRNA | AK033515.1 | - | 2 | 101 | ||
| AK131155.1 | - | 3 | 421 | |||
| AK143714.1 | - | 3 | 441 | |||
| AK159088.1 | - | 3 | 442 | |||
| AK159362.1 | - | 3 | 441 | |||
| AK162424.1 | - | 3 | 443 | |||
| AK172609.1 | - | 3 | 423 | |||
| BB626609.1 | - | 3 | 443 | |||
| BC060617.1 | - | 3 | 426 | 6836791 | ||

Galnt10
Genomic Map