siRNA Details
| Gene Symbol: Fxyd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008557.2 | NP_032583.1 | 5 | 222 | ||
| GenBank mRNA | AA959274.1 | - | 5 | 179 | ||
| AK133713.1 | - | 1 | 719 | |||
| AK145206.1 | - | 193 | ||||
| AK166426.1 | - | 5 | 195 | |||
| BC002039.1 | - | 5 | 186 | 3488180 | ||
| BC051033.1 | - | 5 | 180 | 4460667 | ||
| BC056223.1 | - | 5 | 190 | 6485767 | ||
| X93038.1 | - | 5 | 218 | |||

Fxyd3
Genomic Map