siRNA Details
| Gene Symbol: Meg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK018105.1 | - | 3 | 1347 | ||
| AK141629.1 | - | 3 | 259 | |||
| AK147321.1 | - | 5 | 2814 | |||
| BC024818.1 | - | 5 | 706 | |||
| BC051667.1 | - | 623 | ||||
| BC094421.1 | - | 7 | 1323 | |||
| Y13832.1 | - | LIT3453 Gtl2 |
8 | 1450 | ||

Meg3
Genomic Map