siRNA Details
| Gene Symbol: Neurod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010894.2 | NP_035024.1 | 2 | 219 | ||
| GenBank mRNA | AK005073.1 | - | 2 | 220 | ||
| AK018781.1 | - | 2 | 218 | |||
| AK028055.1 | - | 2 | 223 | |||
| BC018241.1 | - | 2 | 195 | 4511370 | ||
| BC094611.1 | - | 2 | 159 | 30361598 | ||

Neurod1
Genomic Map