siRNA Details
| Gene Symbol: Nkx2-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008701.2 | NP_032727.2 | 1 | 324 | ||
| GenBank mRNA | BC150762.1 | - | 1 | 308 | 9087673 | |
| Y15741.1 | - | 1 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx2-9| Gene Symbol: Nkx2-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008701.2 | NP_032727.2 | 1 | 324 | ||
| GenBank mRNA | BC150762.1 | - | 1 | 308 | 9087673 | |
| Y15741.1 | - | 1 | 325 | |||