siRNA Details
| Gene Symbol: Olfr18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146563.1 | NP_666774.1 | 2 | 196 | ||
| GenBank mRNA | BC120866.1 | - | 2 | 95 | 40129789 | |
| BC120868.1 | - | 2 | 95 | 40129791 | ||
| BC144957.1 | - | 3 | 106 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr18| Gene Symbol: Olfr18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146563.1 | NP_666774.1 | 2 | 196 | ||
| GenBank mRNA | BC120866.1 | - | 2 | 95 | 40129789 | |
| BC120868.1 | - | 2 | 95 | 40129791 | ||
| BC144957.1 | - | 3 | 106 | |||