siRNA Details
| Gene Symbol: Olfr25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146870.2 | NP_667081.2 | 1 | 514 | ||
| GenBank mRNA | BC125306.1 | - | 1 | 514 | 40129821 | |
| BC125308.1 | - | 1 | 514 | 40129823 | ||
| X89684.1 | - | 1 | 34 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr25| Gene Symbol: Olfr25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146870.2 | NP_667081.2 | 1 | 514 | ||
| GenBank mRNA | BC125306.1 | - | 1 | 514 | 40129821 | |
| BC125308.1 | - | 1 | 514 | 40129823 | ||
| X89684.1 | - | 1 | 34 | |||