siRNA Details
Gene Symbol: Olfr32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010980.2 | NP_035110.1 | 5 | 684 | ||
GenBank mRNA | BC119404.1 | - | 1 | 459 | 8734153 | |
BC120653.1 | - | 1 | 459 | 40129576 | ||
X89678.1 | - | 1 | 36 |
