siRNA Details
| Gene Symbol: Ovol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019935.3 | NP_064319.1 | 3 | 672 | ||
| GenBank mRNA | AF134804.1 | - | 3 | 688 | ||
| AF134805.1 | - | 3 | 654 | |||
| AK003801.1 | - | 3 | 707 | |||
| AK028577.1 | - | 3 | 673 | |||
| AK132478.1 | - | 3 | 673 | |||
| AK132505.1 | - | 3 | 711 | |||
| AK132506.1 | - | 3 | 707 | |||
| AK144952.1 | - | 3 | 707 | |||
| BC021411.1 | - | 3 | 752 | 5006712 | ||

Ovol1
Genomic Map