siRNA Details
| Gene Symbol: Pcsk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291184.1 | NP_001278113.1 | 5 | 684 | ||
| NM_011048.1 | NP_035178.1 | 5 | 684 | |||
| GenBank mRNA | BC037450.1 | - | 3 | 289 | ||
| D50060.1 | - | 5 | 607 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcsk6| Gene Symbol: Pcsk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291184.1 | NP_001278113.1 | 5 | 684 | ||
| NM_011048.1 | NP_035178.1 | 5 | 684 | |||
| GenBank mRNA | BC037450.1 | - | 3 | 289 | ||
| D50060.1 | - | 5 | 607 | |||