siRNA Details
| Gene Symbol: Pdgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008808.3 | NP_032834.1 | 5 | 651 | ||
| GenBank mRNA | AK017169.1 | - | 3 | 236 | ||
| AY324648.1 | - | 5 | 640 | |||
| BC003817.1 | - | 5 | 651 | 3495629 | ||
| M29464.1 | - | 5 | 531 | |||

Pdgfa
Genomic Map