siRNA Details
| Gene Symbol: Plk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011121.3 | NP_035251.3 | 8 | 1520 | ||
| GenBank mRNA | AK134045.1 | - | 8 | 1461 | ||
| AK148050.1 | - | 8 | 1434 | |||
| AK148076.1 | - | 8 | 1511 | |||
| AK164027.1 | - | 8 | 1490 | |||
| BC006880.1 | - | 8 | 1472 | 3596381 | ||
| L19558.1 | - | 8 | 1527 | |||
| U01063.1 | - | 8 | 1441 | |||

Plk1
Genomic Map