siRNA Details
| Gene Symbol: Ppp3cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015002.1 | - | 10 | 1349 | ||
| AK133393.1 | - | 10 | 1370 | |||
| BC046617.1 | - | 10 | 1175 | |||
| BC141079.1 | - | 10 | 1190 | 9055691 | ||
| BC145347.1 | - | 9 | 1065 | |||
| M81475.1 | - | 10 | 1343 | |||

Ppp3cc
Genomic Map