siRNA Details
| Gene Symbol: Prkdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011159.2 | NP_035289.2 | 17, 18 | 1920 | ||
| GenBank mRNA | AB007544.1 | - | 17, 18 | 1897 | ||
| AB011543.1 | - | 17, 18 | 1909 | |||
| AK084827.1 | - | 17, 18 | 1921 | |||
| D87521.1 | - | 17, 18 | 1897 | |||

Prkdc
Genomic Map