siRNA Details
| Gene Symbol: CRISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190986.1 | NP_001177915.1 | 8 | 767 | ||
| NM_006061.2 | NP_006052.2 | 8 | 755 | |||
| GenBank mRNA | AK223380.1 | - | 8 | 659 | ||
| AK292786.1 | - | 8 | 714 | |||
| AK303966.1 | - | 6 | 482 | |||
| AK313387.1 | - | 8 | 688 | |||
| BC069580.1 | - | 7 | 641 | 7262396 | ||
| BC069602.1 | - | 7 | 643 | 7262408 | ||
| BC101539.1 | - | 7 | 665 | 8069045 | ||
| X94323.1 | - | 8 | 668 | |||
| X95240.1 | - | 7 | 643 | |||

CRISP3
Genomic Map