siRNA Details
| Gene Symbol: Gm12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195544.1 | NP_001182473.1 | 2 | 539 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12| Gene Symbol: Gm12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195544.1 | NP_001182473.1 | 2 | 539 | ||