siRNA Details
| Gene Symbol: Gm13152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039209.2 | NP_001034298.1 | 793 | |||
| GenBank mRNA | BC058613.1 | - | 793 | 30044792 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13152| Gene Symbol: Gm13152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039209.2 | NP_001034298.1 | 793 | |||
| GenBank mRNA | BC058613.1 | - | 793 | 30044792 | ||